siRNA Details
| Gene Symbol: Ccdc47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013974.1 | NP_001013996.1 | 12 | 1422 | ||
| GenBank mRNA | BC085824.1 | - | 12 | 1422 | 7127239 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc47| Gene Symbol: Ccdc47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013974.1 | NP_001013996.1 | 12 | 1422 | ||
| GenBank mRNA | BC085824.1 | - | 12 | 1422 | 7127239 | |