siRNA Details
| Gene Symbol: Ftsj3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012014.1 | NP_001012014.1 | 9, 10 | 876 | ||
| GenBank mRNA | BC086512.1 | - | 9, 10 | 876 | 7133710 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ftsj3| Gene Symbol: Ftsj3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012014.1 | NP_001012014.1 | 9, 10 | 876 | ||
| GenBank mRNA | BC086512.1 | - | 9, 10 | 876 | 7133710 | |