siRNA Details
| Gene Symbol: Tex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247645.1 | XP_006247707.1 | 2 | 1108 | ||
| XM_006247646.2 | XP_006247708.1 | 2 | 1940 | |||
| XM_006247647.1 | XP_006247709.1 | 2 | 1108 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tex2| Gene Symbol: Tex2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006247645.1 | XP_006247707.1 | 2 | 1108 | ||
| XM_006247646.2 | XP_006247708.1 | 2 | 1940 | |||
| XM_006247647.1 | XP_006247709.1 | 2 | 1108 | |||