siRNA Details
Gene Symbol: Bptf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768403.1 | XP_008766625.1 | 29 | 8807 | ||
GenBank mRNA | BC160926.1 | - | 3 | 672 | 8364269 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Bptf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008768403.1 | XP_008766625.1 | 29 | 8807 | ||
GenBank mRNA | BC160926.1 | - | 3 | 672 | 8364269 |