siRNA Details
Gene Symbol: Cog1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107062.2 | NP_001100532.2 | 7 | 1559 | ||
XM_006247686.2 | XP_006247748.1 | 7 | 1828 | |||
XM_006247687.2 | XP_006247749.1 | 7 | 1828 | |||
GenBank mRNA | BC169068.1 | - | 1557 | |||
FQ212043.1 | - | 7 | 1833 |
