siRNA Details
| Gene Symbol: Cog1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107062.2 | NP_001100532.2 | 7 | 1377 | ||
| XM_006247686.2 | XP_006247748.1 | 7 | 1646 | |||
| XM_006247687.2 | XP_006247749.1 | 7 | 1646 | |||
| GenBank mRNA | BC169068.1 | - | 1375 | |||
| FQ212043.1 | - | 7 | 1651 | |||

Cog1
Genomic Map