siRNA Details
| Gene Symbol: Engase | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107070.1 | NP_001100540.1 | 12, 13 | 1825 | ||
| XM_006247803.2 | XP_006247865.1 | 12, 13 | 1815 | |||
| XM_006247804.2 | XP_006247866.1 | 12, 13 | 1640 | |||
| GenBank mRNA | FQ212026.1 | - | 4, 5 | 671 | ||

Engase
Genomic Map