siRNA Details
| Gene Symbol: Arvcf | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001131013.1 | NP_001124485.1 | 15 | 2829 | ||
| XM_006248617.2 | XP_006248679.1 | 14 | 5079 | |||
| XM_006248618.2 | XP_006248680.1 | 13 | 5061 | |||
| XM_008768880.1 | XP_008767102.1 | 16 | 2838 | |||
| XM_008768881.1 | XP_008767103.1 | 15 | 3802 | |||
| XM_008768882.1 | XP_008767104.1 | 14 | 2948 | |||
| XM_008768883.1 | XP_008767105.1 | 15 | 3142 | |||
| XM_008768884.1 | XP_008767106.1 | 14 | 3123 | |||
| XM_008768885.1 | XP_008767107.1 | 14 | 5079 | |||
| GenBank mRNA | BC168253.1 | - | 15 | 2829 | 5621230 | |

Arvcf
Genomic Map