siRNA Details
| Gene Symbol: Tbccd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012016.1 | NP_001012016.1 | 3 | 555 | ||
| XM_006248545.2 | XP_006248607.1 | 2 | 529 | |||
| XM_006248546.2 | XP_006248608.1 | 2 | 677 | |||
| GenBank mRNA | BC079273.1 | - | 3 | 585 | ||
| BC089823.1 | - | 3 | 555 | 7133137 | ||

Tbccd1
Genomic Map