siRNA Details
| Gene Symbol: Tbccd1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012016.1 | NP_001012016.1 | 5 | 1063 | ||
| XM_006248545.2 | XP_006248607.1 | 4 | 1037 | |||
| XM_006248546.2 | XP_006248608.1 | 4 | 1185 | |||
| GenBank mRNA | BC079273.1 | - | 5 | 1093 | ||
| BC089823.1 | - | 5 | 1063 | 7133137 | ||

Tbccd1
Genomic Map