siRNA Details
| Gene Symbol: Muc4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006248471.2 | XP_006248533.1 | 21, 22 | 6293 | ||
| XM_008768775.1 | XP_008766997.1 | 21, 22 | 5933 | |||
| XM_008768776.1 | XP_008766998.1 | 21, 22 | 5201 | |||
| XM_008768777.1 | XP_008766999.1 | 21, 22 | 4470 | |||
| GenBank mRNA | U06752.1 | - | 22, 23 | 4674 | ||

Muc4
Genomic Map