siRNA Details
| Gene Symbol: Parp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191659.1 | NP_001178588.1 | 14 | 4731 | ||
| GenBank mRNA | FQ218194.1 | - | 7 | 1350 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Parp14| Gene Symbol: Parp14 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191659.1 | NP_001178588.1 | 14 | 4731 | ||
| GenBank mRNA | FQ218194.1 | - | 7 | 1350 | ||