siRNA Details
| Gene Symbol: Iqcb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107092.1 | NP_001100562.1 | 5 | 440 | ||
| XM_006248395.2 | XP_006248457.1 | 5 | 506 | |||
| XM_006248396.2 | XP_006248458.1 | 5 | 511 | |||
| XM_006248397.2 | XP_006248459.1 | 5 | 474 | |||
| XM_008768730.1 | XP_008766952.1 | 5 | 479 | |||

Iqcb1
Genomic Map