siRNA Details
Gene Symbol: Iqcb1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107092.1 | NP_001100562.1 | 10 | 1014 | ||
XM_006248395.2 | XP_006248457.1 | 10 | 1080 | |||
XM_006248396.2 | XP_006248458.1 | 10 | 1085 | |||
XM_006248397.2 | XP_006248459.1 | 10 | 1048 | |||
XM_008768730.1 | XP_008766952.1 | 10 | 1053 |
