siRNA Details
| Gene Symbol: Gtf2e1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001100556.1 | NP_001094026.1 | 5 | 1417 | ||
| XM_006248398.2 | XP_006248460.1 | 5 | 1747 | |||
| XM_008768731.1 | XP_008766953.1 | 5 | 2269 | |||
| GenBank mRNA | BC099237.1 | - | 4 | 956 | ||

Gtf2e1
Genomic Map