siRNA Details
| Gene Symbol: Timmdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007676.1 | NP_001007677.1 | 3 | 527 | ||
| GenBank mRNA | BC079140.1 | - | 3 | 527 | 7127856 | |
| FQ215140.1 | - | 3 | 554 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Timmdc1| Gene Symbol: Timmdc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001007676.1 | NP_001007677.1 | 3 | 527 | ||
| GenBank mRNA | BC079140.1 | - | 3 | 527 | 7127856 | |
| FQ215140.1 | - | 3 | 554 | |||