siRNA Details
| Gene Symbol: Igsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013120.1 | NP_001013138.1 | 7 | 1211 | ||
| GenBank mRNA | BC085929.1 | - | 7 | 1211 | 7110445 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Igsf11| Gene Symbol: Igsf11 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001013120.1 | NP_001013138.1 | 7 | 1211 | ||
| GenBank mRNA | BC085929.1 | - | 7 | 1211 | 7110445 | |