siRNA Details
Gene Symbol: Nfkbiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107095.1 | NP_001100565.1 | 9, 10 | 1618 | ||
GenBank mRNA | AB438101.1 | - | 12, 13 | 2142 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Nfkbiz | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107095.1 | NP_001100565.1 | 9, 10 | 1618 | ||
GenBank mRNA | AB438101.1 | - | 12, 13 | 2142 |