siRNA Details
| Gene Symbol: Trmt10c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008337.1 | NP_001008338.1 | 2 | 420 | ||
| GenBank mRNA | BC085895.1 | - | 2 | 420 | 7193162 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Trmt10c| Gene Symbol: Trmt10c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001008337.1 | NP_001008338.1 | 2 | 420 | ||
| GenBank mRNA | BC085895.1 | - | 2 | 420 | 7193162 | |