siRNA Details
| Gene Symbol: LOC100911145 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751038.3 | XP_003751086.1 | 9 | 1065 | ||
| Gene Symbol: Gpr128 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107098.1 | NP_001100568.1 | 1237 | |||
More Information
Set Membership: |
|

LOC100911145
Genomic Map