siRNA Details
| Gene Symbol: Col8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107100.1 | NP_001100570.1 | 3 | 434 | ||
| XM_006248207.1 | XP_006248269.1 | 1 | 240 | |||
| XM_006248208.2 | XP_006248270.1 | 1 | 239 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Col8a1| Gene Symbol: Col8a1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107100.1 | NP_001100570.1 | 3 | 434 | ||
| XM_006248207.1 | XP_006248269.1 | 1 | 240 | |||
| XM_006248208.2 | XP_006248270.1 | 1 | 239 | |||