siRNA Details
| Gene Symbol: Fam3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107102.1 | NP_001100572.1 | 6 | 512 | ||
| XM_006248157.2 | XP_006248219.1 | 6 | 575 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam3b| Gene Symbol: Fam3b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107102.1 | NP_001100572.1 | 6 | 512 | ||
| XM_006248157.2 | XP_006248219.1 | 6 | 575 | |||