siRNA Details
| Gene Symbol: Dopey2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191660.1 | NP_001178589.1 | 20 | 4938 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Dopey2| Gene Symbol: Dopey2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191660.1 | NP_001178589.1 | 20 | 4938 | ||