siRNA Details
| Gene Symbol: Clic6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176078.2 | NP_788267.1 | 2 | 1483 | ||
| GenBank mRNA | AY191249.1 | - | 2 | 1483 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Clic6| Gene Symbol: Clic6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_176078.2 | NP_788267.1 | 2 | 1483 | ||
| GenBank mRNA | AY191249.1 | - | 2 | 1483 | ||