siRNA Details
| Gene Symbol: Son | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001170327.1 | NP_001163798.1 | 3 | 3717 | ||
| NM_001170328.1 | NP_001163799.1 | 3 | 3717 | |||
| XM_006248064.2 | XP_006248126.1 | 3 | 3735 | |||
| XM_006248065.2 | XP_006248127.1 | 3 | 3735 | |||
| XM_008768580.1 | XP_008766802.1 | 3 | 3735 | |||

Son
Genomic Map