siRNA Details
| Gene Symbol: Bach1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107113.1 | NP_001100583.1 | 3 | 1513 | ||
| XM_003751009.3 | XP_003751057.2 | 3 | 1558 | |||
| XM_006248018.2 | XP_006248080.1 | 3 | 1558 | |||
| XM_006248019.2 | XP_006248081.1 | 3 | 2717 | |||
| XM_006248020.2 | XP_006248082.1 | 3 | 2574 | |||
| XM_008768581.1 | XP_008766803.1 | 3 | 3317 | |||

Bach1
Genomic Map