siRNA Details
| Gene Symbol: Zkscan5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107119.1 | NP_001100589.1 | 3 | 1249 | ||
| GenBank mRNA | BC167019.1 | - | 3 | 1165 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Zkscan5| Gene Symbol: Zkscan5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107119.1 | NP_001100589.1 | 3 | 1249 | ||
| GenBank mRNA | BC167019.1 | - | 3 | 1165 | ||