siRNA Details
Gene Symbol: Ptcd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001109665.1 | NP_001103135.1 | 5 | 1523 | ||
XM_006248871.1 | XP_006248933.1 | 6 | 1654 | |||
XM_006248873.1 | XP_006248935.1 | 5 | 1513 | |||
GenBank mRNA | BC129116.1 | - | 6 | 1563 | 7113830 |
