siRNA Details
| Gene Symbol: Zfp316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107121.1 | NP_001100591.1 | 2 | 478 | ||
| XM_008768963.1 | XP_008767185.1 | 2 | 2900 | |||
| XM_008768964.1 | XP_008767186.1 | 3 | 2551 | |||
| XM_008768965.1 | XP_008767187.1 | 2 | 2434 | |||
| XM_008768966.1 | XP_008767188.1 | 3 | 2491 | |||
| XM_008768967.1 | XP_008767189.1 | 2 | 2022 | |||
| XM_008768968.1 | XP_008767190.1 | 2 | 466 | |||

Zfp316
Genomic Map