siRNA Details
| Gene Symbol: Zfp316 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107121.1 | NP_001100591.1 | 6 | 1912 | ||
| XM_008768963.1 | XP_008767185.1 | 6 | 4334 | |||
| XM_008768964.1 | XP_008767186.1 | 7 | 3985 | |||
| XM_008768965.1 | XP_008767187.1 | 6 | 3868 | |||
| XM_008768966.1 | XP_008767188.1 | 7 | 3925 | |||
| XM_008768967.1 | XP_008767189.1 | 6 | 3456 | |||
| XM_008768968.1 | XP_008767190.1 | 6 | 1900 | |||

Zfp316
Genomic Map