siRNA Details
| Gene Symbol: Fam20c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012238.1 | NP_001012238.1 | 2 | 1207 | ||
| GenBank mRNA | BC161825.1 | - | 2 | 874 | 8373563 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Fam20c| Gene Symbol: Fam20c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012238.1 | NP_001012238.1 | 2 | 1207 | ||
| GenBank mRNA | BC161825.1 | - | 2 | 874 | 8373563 | |