siRNA Details
| Gene Symbol: Zscan21 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001012021.2 | NP_001012021.1 | 2 | 461 | ||
| XM_006249007.2 | XP_006249069.1 | 2 | 590 | |||
| GenBank mRNA | BC079435.1 | - | 2 | 409 | 7115285 | |
| FM121830.1 | - | 2 | 461 | |||

Zscan21
Genomic Map