siRNA Details
Gene Symbol: Tyw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107137.1 | NP_001100607.1 | 16 | 2294 | ||
GenBank mRNA | BC168924.1 | - | 16 | 2276 | 9029270 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Tyw1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107137.1 | NP_001100607.1 | 16 | 2294 | ||
GenBank mRNA | BC168924.1 | - | 16 | 2276 | 9029270 |