siRNA Details
| Gene Symbol: Sbno1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107138.1 | NP_001100608.1 | 21, 22 | 2986 | ||
| XM_006249300.2 | XP_006249362.1 | 21, 22 | 3563 | |||
| XM_006249301.2 | XP_006249363.1 | 21, 22 | 3560 | |||
| XM_008769224.1 | XP_008767446.1 | 20, 21 | 3458 | |||
| XM_008769225.1 | XP_008767447.1 | 20, 21 | 3455 | |||
| XM_008769226.1 | XP_008767448.1 | 18, 19 | 2468 | |||

Sbno1
Genomic Map