siRNA Details
| Gene Symbol: Tctn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177613.1 | NP_001171084.1 | 12 | 1408 | ||
| GenBank mRNA | FQ222752.1 | - | 11 | 1488 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tctn1| Gene Symbol: Tctn1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001177613.1 | NP_001171084.1 | 12 | 1408 | ||
| GenBank mRNA | FQ222752.1 | - | 11 | 1488 | ||