siRNA Details
Gene Symbol: Anapc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107142.1 | NP_001100612.1 | 5, 6 | 796 | ||
XM_006249365.1 | XP_006249427.1 | 4, 5 | 714 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Anapc7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107142.1 | NP_001100612.1 | 5, 6 | 796 | ||
XM_006249365.1 | XP_006249427.1 | 4, 5 | 714 |