siRNA Details
Gene Symbol: LOC100359816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769303.1 | XP_008767525.1 | 18 | 6117 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC100359816 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | XM_008769303.1 | XP_008767525.1 | 18 | 6117 |