siRNA Details
Gene Symbol: Rfc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107146.2 | NP_001100616.1 | 6, 7 | 664 | ||
GenBank mRNA | BC168902.1 | - | 6, 7 | 664 | 7388155 |

Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: Rfc5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001107146.2 | NP_001100616.1 | 6, 7 | 664 | ||
GenBank mRNA | BC168902.1 | - | 6, 7 | 664 | 7388155 |