siRNA Details
| Gene Symbol: Tchp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191666.1 | NP_001178595.1 | 8 | 925 | ||
| XM_006249451.1 | XP_006249513.1 | 9 | 951 | |||
| XM_006249452.1 | XP_006249514.1 | 8 | 938 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Tchp| Gene Symbol: Tchp | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001191666.1 | NP_001178595.1 | 8 | 925 | ||
| XM_006249451.1 | XP_006249513.1 | 9 | 951 | |||
| XM_006249452.1 | XP_006249514.1 | 8 | 938 | |||