siRNA Details
| Gene Symbol: Ep400 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107149.1 | NP_001100619.1 | 8, 9 | 2682 | ||
| XM_006249549.2 | XP_006249611.1 | 9, 10 | 2793 | |||
| XM_006249550.2 | XP_006249612.1 | 9, 10 | 2958 | |||
| XM_006249552.2 | XP_006249614.1 | 9, 10 | 2774 | |||
| XM_006249553.2 | XP_006249615.1 | 9, 10 | 2794 | |||
| XM_006249554.2 | XP_006249616.1 | 9, 10 | 2793 | |||
| XM_006249556.2 | XP_006249618.1 | 8, 9 | 2685 | |||
| XM_006249557.2 | XP_006249619.1 | 8, 9 | 2682 | |||
| XM_006249558.2 | XP_006249620.1 | 9, 10 | 2793 | |||
| XM_006249559.2 | XP_006249621.1 | 7, 8 | 2573 | |||
| XM_006249560.2 | XP_006249622.1 | 8, 9 | 2681 | |||
| XM_008769332.1 | XP_008767554.1 | 10, 11 | 2895 | |||

Ep400
Genomic Map