siRNA Details
| Gene Symbol: Baz2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107158.1 | NP_001100628.1 | 6 | 1214 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Baz2a| Gene Symbol: Baz2a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107158.1 | NP_001100628.1 | 6 | 1214 | ||