siRNA Details
| Gene Symbol: Ikzf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240732.2 | XP_006240794.1 | 8 | 1590 | ||
| XM_008765052.1 | XP_008763274.1 | 7 | 1475 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ikzf4| Gene Symbol: Ikzf4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_006240732.2 | XP_006240794.1 | 8 | 1590 | ||
| XM_008765052.1 | XP_008763274.1 | 7 | 1475 | |||