siRNA Details
| Gene Symbol: Serpinb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135867.1 | NP_001129339.1 | 8 | 865 | ||
| XM_006249637.2 | XP_006249699.1 | 7 | 877 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Serpinb12| Gene Symbol: Serpinb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135867.1 | NP_001129339.1 | 8 | 865 | ||
| XM_006249637.2 | XP_006249699.1 | 7 | 877 | |||