siRNA Details
| Gene Symbol: Serpinb12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135867.1 | NP_001129339.1 | 6 | 648 | ||
| XM_006249637.2 | XP_006249699.1 | 5 | 660 | |||
| Gene Symbol: LOC103690041 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | XM_003751227.3 | XP_003751275.3 | 4 | 550 | ||

Serpinb12
Genomic Map