siRNA Details
| Gene Symbol: Rrn3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001135846.1 | NP_001129318.1 | 13 | 1258 | ||
| XM_008767458.1 | XP_008765680.1 | 13 | 1282 | |||
| XM_008767459.1 | XP_008765681.1 | 13 | 1633 | |||
| GenBank mRNA | BC166485.1 | - | 13 | 1258 | 7099831 | |
| FQ228986.1 | - | 13 | 1278 | |||

Rrn3
Genomic Map