siRNA Details
| Gene Symbol: Ccdc93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024997.1 | NP_001020168.1 | 8 | 771 | ||
| XM_008769456.1 | XP_008767678.1 | 9 | 805 | |||
| GenBank mRNA | BC091337.1 | - | 8 | 771 | 7314934 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ccdc93| Gene Symbol: Ccdc93 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024997.1 | NP_001020168.1 | 8 | 771 | ||
| XM_008769456.1 | XP_008767678.1 | 9 | 805 | |||
| GenBank mRNA | BC091337.1 | - | 8 | 771 | 7314934 | |