siRNA Details
| Gene Symbol: Ccnt2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107171.1 | NP_001100641.1 | 9 | 1868 | ||
| XM_006249686.2 | XP_006249748.1 | 9 | 1876 | |||
| XM_006249687.2 | XP_006249749.1 | 5 | 4577 | |||
| XM_008769457.1 | XP_008767679.1 | 8 | 1747 | |||
| XM_008769458.1 | XP_008767680.1 | 6 | 3653 | |||

Ccnt2
Genomic Map