siRNA Details
| Gene Symbol: Lmod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107179.1 | NP_001100649.1 | 1 | 361 | ||
| XM_008769562.1 | XP_008767784.1 | 1 | 789 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Lmod1| Gene Symbol: Lmod1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001107179.1 | NP_001100649.1 | 1 | 361 | ||
| XM_008769562.1 | XP_008767784.1 | 1 | 789 | |||