siRNA Details
| Gene Symbol: Cdc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024769.1 | NP_001019940.1 | 14 | 1320 | ||
| GenBank mRNA | BC097445.1 | - | 14 | 1320 | 7456471 | |
Please enter the information below and press OK to send your cart to Core Services for purchase.

Cdc73| Gene Symbol: Cdc73 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001024769.1 | NP_001019940.1 | 14 | 1320 | ||
| GenBank mRNA | BC097445.1 | - | 14 | 1320 | 7456471 | |