siRNA Details
| Gene Symbol: Trmt1l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001037192.1 | NP_001032269.1 | 5 | 796 | ||
| XM_006249992.2 | XP_006250054.1 | 5 | 894 | |||
| XM_006249993.2 | XP_006250055.1 | 5 | 899 | |||
| XM_008769627.1 | XP_008767849.1 | 6 | 1240 | |||
| XM_008769628.1 | XP_008767850.1 | 3 | 460 | |||
| XM_008769629.1 | XP_008767851.1 | 3 | 423 | |||
| XM_008769630.1 | XP_008767852.1 | 3 | 423 | |||
| XM_008769631.1 | XP_008767853.1 | 4 | 717 | |||
| XM_008769632.1 | XP_008767854.1 | 4 | 338 | |||
| GenBank mRNA | BC100650.1 | - | 5 | 796 | 7932888 | |

Trmt1l
Genomic Map